3-Chlor-3',5'-dimethyl-4'-methoxybenzophenon

Lieferant: FLUOROCHEM

022721-1G
FLUO022721-1GEA 882 EUR
FLUO022721-1G
3-Chlor-3',5'-dimethyl-4'-methoxybenzophenon
3-Chlor-3',5'-dimethyl-4'-methoxybenzophenon
Formel: C₁₆H₁₅ClO₂
MDL: MFCD06201490
CAS-Nummer: 61259-85-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR