3-Chlor-3',4'-dimethylbenzophenon

Lieferant: FLUOROCHEM

022722-1G
FLUO022722-1GEA 794 EUR
FLUO022722-1G
3-Chlor-3',4'-dimethylbenzophenon
3-Chlor-3',4'-dimethylbenzophenon
Formel: C₁₅H₁₃ClO
MDL: MFCD06201491
CAS-Nummer: 844884-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR