3-Chlor-4'-(ethylthio)benzophenon

Lieferant: FLUOROCHEM

022724-1G
FLUO022724-1GEA 882 EUR
FLUO022724-1G
3-Chlor-4'-(ethylthio)benzophenon
3-Chlor-4'-(ethylthio)benzophenon
Formel: C₁₅H₁₃ClOS
MDL: MFCD06201493
CAS-Nummer: 844884-99-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR