3-Chlor-3'-fluor-4'-methoxybenzophenon
Lieferant: FLUOROCHEM
022725-1G
FLUO022725-1GEA
830
EUR
FLUO022725-1G
3-Chlor-3'-fluor-4'-methoxybenzophenon
3-Chlor-3'-fluor-4'-methoxybenzophenon
Formel:
C₁₄H₁₀ClFO₂ |
MDL:
MFCD06201494 CAS-Nummer: 844885-00-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...