4-Chlor-4'-methylbenzophenon
Lieferant: FLUOROCHEM
Synonyme:
(4-Chlorophenyl)(p-tolyl)methanone
022727-1G
FLUO022727-1GEA
461
EUR
FLUO022727-1G
4-Chlor-4'-methylbenzophenon
4-Chlor-4'-methylbenzophenon
Formel:
C₁₄H₁₁ClO Molecular Weight: 230.690 g/mol |
MDL:
MFCD00625699 CAS-Nummer: 5395-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...