4-Chlor-3'-methoxybenzophenon

Lieferant: FLUOROCHEM

022728-1G
FLUO022728-1GEA 812 EUR
FLUO022728-1G
4-Chlor-3'-methoxybenzophenon
4-Chlor-3'-methoxybenzophenon
Formel: C₁₄H₁₁ClO₂
MDL: MFCD01311567
CAS-Nummer: 32363-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR