4-Chlor-4'-methoxybenzophenon

Lieferant: FLUOROCHEM

022729-1G
FLUO022729-1GEA 116 EUR
FLUO022729-1G
4-Chlor-4'-methoxybenzophenon
4-Chlor-4'-methoxybenzophenon
Formel: C₁₄H₁₁ClO₂
MDL: MFCD01105071
CAS-Nummer: 10547-60-1
EINECS: 234-131-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR