4-Chlor-4'-(methylthio)benzophenon

Lieferant: FLUOROCHEM

022730-1G
FLUO022730-1GEA 830 EUR
FLUO022730-1G
4-Chlor-4'-(methylthio)benzophenon
4-Chlor-4'-(methylthio)benzophenon
Formel: C₁₄H₁₁ClOS
MDL: MFCD06201496
CAS-Nummer: 72585-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR