4-tert-Butyl-4'-chlorbenzophenon

Lieferant: FLUOROCHEM

022732-1G
FLUO022732-1GEA 882 EUR
FLUO022732-1G
4-tert-Butyl-4'-chlorbenzophenon
4-tert-Butyl-4'-chlorbenzophenon
Formel: C₁₇H₁₇ClO
MDL: MFCD06201498
CAS-Nummer: 67743-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR