4-Butyl-4'-chlorbenzophenon

Lieferant: FLUOROCHEM

022735-1G
FLUO022735-1GEA 882 EUR
FLUO022735-1G
4-Butyl-4'-chlorbenzophenon
4-Butyl-4'-chlorbenzophenon
Formel: C₁₇H₁₇ClO
MDL: MFCD06201501
CAS-Nummer: 64357-64-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR