4-Chlor-3',4'-difluorbenzophenon

Lieferant: FLUOROCHEM

022736-1G
FLUO022736-1GEA 830 EUR
FLUO022736-1G
4-Chlor-3',4'-difluorbenzophenon
4-Chlor-3',4'-difluorbenzophenon
Formel: C₁₃H₇ClF₂O
MDL: MFCD06201502
CAS-Nummer: 844885-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR