3,4'-Dichlor-5-fluorbenzophenon
Lieferant: FLUOROCHEM
022739-1G
FLUO022739-1GEA
830
EUR
FLUO022739-1G
3,4'-Dichlor-5-fluorbenzophenon
3,4'-Dichlor-5-fluorbenzophenon
Formel:
C₁₃H₇Cl₂FO |
MDL:
MFCD06201505 CAS-Nummer: 844885-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...