4-Chlor-3',5'-dimethyl-4'-methoxybenzophenon

Lieferant: FLUOROCHEM

Synonyme: (4-Chlorophenyl)(4-methoxy-3,5-dimethylphenyl)methanone

022741-1G
FLUO022741-1GEA 882 EUR
FLUO022741-1G
4-Chlor-3',5'-dimethyl-4'-methoxybenzophenon
4-Chlor-3',5'-dimethyl-4'-methoxybenzophenon
Formel: C₁₆H₁₅ClO₂
Molecular Weight: 274,742 g/mol
MDL: MFCD06201506
CAS-Nummer: 61259-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR