4-Chlor-3',4'-dimethylbenzophenon

Lieferant: FLUOROCHEM

022742-1G
FLUO022742-1GEA 830 EUR
FLUO022742-1G
4-Chlor-3',4'-dimethylbenzophenon
4-Chlor-3',4'-dimethylbenzophenon
Formel: C₁₅H₁₃ClO
MDL: MFCD06201507
CAS-Nummer: 40415-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR