4-Chlor-3',5'-dimethylbenzophenon

Lieferant: FLUOROCHEM

022743-1G
FLUO022743-1GEA 830 EUR
FLUO022743-1G
4-Chlor-3',5'-dimethylbenzophenon
4-Chlor-3',5'-dimethylbenzophenon
Formel: C₁₅H₁₃ClO
MDL: MFCD06201508
CAS-Nummer: 844885-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR