4-Chlor-3'-fluor-4'-methoxybenzophenon
Lieferant: FLUOROCHEM
022745-1G
FLUO022745-1GEA
882
EUR
FLUO022745-1G
4-Chlor-3'-fluor-4'-methoxybenzophenon
4-Chlor-3'-fluor-4'-methoxybenzophenon
Formel:
C₁₄H₁₀ClFO₂ |
MDL:
MFCD06201510 CAS-Nummer: 844885-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...