3,5-Difluor-3'-methoxybenzophenon

Lieferant: FLUOROCHEM

022748-1G
FLUO022748-1GEA 830 EUR
FLUO022748-1G
3,5-Difluor-3'-methoxybenzophenon
3,5-Difluor-3'-methoxybenzophenon
Formel: C₁₄H₁₀F₂O₂
MDL: MFCD06201513
CAS-Nummer: 844885-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR