3,5-Difluor-4'-methoxybenzophenon

Lieferant: FLUOROCHEM

022749-1G
FLUO022749-1GEA 830 EUR
FLUO022749-1G
3,5-Difluor-4'-methoxybenzophenon
3,5-Difluor-4'-methoxybenzophenon
Formel: C₁₄H₁₀F₂O₂
MDL: MFCD06201514
CAS-Nummer: 148560-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR