3,5-Difluor-4'-(methylthio)benzophenon
Lieferant: FLUOROCHEM
022750-1G
FLUO022750-1GEA
830
EUR
FLUO022750-1G
3,5-Difluor-4'-(methylthio)benzophenon
3,5-Difluor-4'-(methylthio)benzophenon
Formel:
C₁₄H₁₀F₂OS |
MDL:
MFCD06201515 CAS-Nummer: 3411-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...