4-Methoxy-3,3',5'-trifluorbenzophenon
Lieferant: FLUOROCHEM
022756-1G
FLUO022756-1GEA
830
EUR
FLUO022756-1G
4-Methoxy-3,3',5'-trifluorbenzophenon
4-Methoxy-3,3',5'-trifluorbenzophenon
Formel:
C₁₄H₉F₃O₂ |
MDL:
MFCD06201521 CAS-Nummer: 844885-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...