3,4-Dichlor-3',5'-difluorbenzophenon

Lieferant: FLUOROCHEM

022759-1G
FLUO022759-1GEA 830 EUR
FLUO022759-1G
3,4-Dichlor-3',5'-difluorbenzophenon
3,4-Dichlor-3',5'-difluorbenzophenon
Formel: C₁₃H₆Cl₂F₂O
MDL: MFCD06201524
CAS-Nummer: 844885-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR