3,5-Dichlor-3',5'-difluorbenzophenon

Lieferant: FLUOROCHEM

022760-1G
FLUO022760-1GEA 830 EUR
FLUO022760-1G
3,5-Dichlor-3',5'-difluorbenzophenon
3,5-Dichlor-3',5'-difluorbenzophenon
Formel: C₁₃H₆Cl₂F₂O
MDL: MFCD06201525
CAS-Nummer: 844885-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR