3-Chlor-3',5,5'-trifluorbenzophenon
Lieferant: FLUOROCHEM
022761-1G
FLUO022761-1GEA
830
EUR
FLUO022761-1G
3-Chlor-3',5,5'-trifluorbenzophenon
3-Chlor-3',5,5'-trifluorbenzophenon
Formel:
C₁₃H₆ClF₃O |
MDL:
MFCD06201526 CAS-Nummer: 844885-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...