3,5-Difluor-3',4'-dimethylbenzophenon

Lieferant: FLUOROCHEM

022764-1G
FLUO022764-1GEA 830 EUR
FLUO022764-1G
3,5-Difluor-3',4'-dimethylbenzophenon
3,5-Difluor-3',4'-dimethylbenzophenon
Formel: C₁₅H₁₂F₂O
MDL: MFCD06201529
CAS-Nummer: 844885-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR