3,5-Difluor-3',5'-dimethylbenzophenon
Lieferant: FLUOROCHEM
022765-1G
FLUO022765-1GEA
1040
EUR
FLUO022765-1G
3,5-Difluor-3',5'-dimethylbenzophenon
3,5-Difluor-3',5'-dimethylbenzophenon
Formel:
C₁₅H₁₂F₂O |
MDL:
MFCD06201530 CAS-Nummer: 844885-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...