3,5-Difluor-4'-(ethylthio)benzophenon

Lieferant: FLUOROCHEM

022766-1G
FLUO022766-1GEA 1040 EUR
FLUO022766-1G
3,5-Difluor-4'-(ethylthio)benzophenon
3,5-Difluor-4'-(ethylthio)benzophenon
Formel: C₁₅H₁₂F₂OS
MDL: MFCD06201531
CAS-Nummer: 844885-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR