3,4-Dichlor-3'-methoxybenzophenon
Lieferant: FLUOROCHEM
022769-1G
FLUO022769-1GEA
1100
EUR
FLUO022769-1G
3,4-Dichlor-3'-methoxybenzophenon
3,4-Dichlor-3'-methoxybenzophenon
Formel:
C₁₄H₁₀Cl₂O₂ |
MDL:
MFCD06201534 CAS-Nummer: 844885-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...