3,4-Dichlor-4'-methoxybenzophenon
Lieferant: FLUOROCHEM
022770-1G
FLUO022770-1GEA
830
EUR
FLUO022770-1G
3,4-Dichlor-4'-methoxybenzophenon
3,4-Dichlor-4'-methoxybenzophenon
Formel:
C₁₄H₁₀Cl₂O₂ |
MDL:
MFCD06201535 CAS-Nummer: 66938-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...