3,4-Dichlor-4'-(methylthio)benzophenon
Lieferant: FLUOROCHEM
022771-1G
FLUO022771-1GEA
830
EUR
FLUO022771-1G
3,4-Dichlor-4'-(methylthio)benzophenon
3,4-Dichlor-4'-(methylthio)benzophenon
Formel:
C₁₄H₁₀Cl₂OS |
MDL:
MFCD06201536 CAS-Nummer: 73242-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...