4-tert-Butyl-3',4'-dichlorbenzophenon

Lieferant: FLUOROCHEM

022773-1G
FLUO022773-1GEA 882 EUR
FLUO022773-1G
4-tert-Butyl-3',4'-dichlorbenzophenon
4-tert-Butyl-3',4'-dichlorbenzophenon
Formel: C₁₇H₁₆Cl₂O
MDL: MFCD06201538
CAS-Nummer: 844885-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR