3,4-Dichlor-4'-propylbenzophenon

Lieferant: FLUOROCHEM

022775-1G
FLUO022775-1GEA 882 EUR
FLUO022775-1G
3,4-Dichlor-4'-propylbenzophenon
3,4-Dichlor-4'-propylbenzophenon
Formel: C₁₆H₁₄Cl₂O
MDL: MFCD06201540
CAS-Nummer: 844885-29-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR