3,4-Dichlor-3',4'-difluorbenzophenon
Lieferant: FLUOROCHEM
022777-1G
FLUO022777-1GEA
830
EUR
FLUO022777-1G
3,4-Dichlor-3',4'-difluorbenzophenon
3,4-Dichlor-3',4'-difluorbenzophenon
Formel:
C₁₃H₆Cl₂F₂O |
MDL:
MFCD06201542 CAS-Nummer: 844885-31-0 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...