3,4-Dichlor-3',4'-dimethoxybenzophenon

Lieferant: FLUOROCHEM

022781-1G
FLUO022781-1GEA 830 EUR
FLUO022781-1G
3,4-Dichlor-3',4'-dimethoxybenzophenon
3,4-Dichlor-3',4'-dimethoxybenzophenon
Formel: C₁₅H₁₂Cl₂O₃
MDL: MFCD06201546
CAS-Nummer: 92436-56-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR