3,4-Dichlor-3',4'-dimethylbenzophenon

Lieferant: FLUOROCHEM

022783-1G
FLUO022783-1GEA 882 EUR
FLUO022783-1G
3,4-Dichlor-3',4'-dimethylbenzophenon
3,4-Dichlor-3',4'-dimethylbenzophenon
Formel: C₁₅H₁₂Cl₂O
MDL: MFCD06201548
CAS-Nummer: 844885-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR