3,4-Dichlor-3',5'-dimethylbenzophenon
Lieferant: FLUOROCHEM
022784-1G
FLUO022784-1GEA
849
EUR
FLUO022784-1G
3,4-Dichlor-3',5'-dimethylbenzophenon
3,4-Dichlor-3',5'-dimethylbenzophenon
Formel:
C₁₅H₁₂Cl₂O |
MDL:
MFCD06201549 CAS-Nummer: 844885-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...