3,4-Dichlor-4'-(ethylthio)benzophenon

Lieferant: FLUOROCHEM

022785-1G
FLUO022785-1GEA 882 EUR
FLUO022785-1G
3,4-Dichlor-4'-(ethylthio)benzophenon
3,4-Dichlor-4'-(ethylthio)benzophenon
Formel: C₁₅H₁₂Cl₂OS
MDL: MFCD06201550
CAS-Nummer: 844885-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR