3,4-Dichlor-3'-fluor-4'-methoxybenzophenon

Lieferant: FLUOROCHEM

022786-1G
FLUO022786-1GEA 830 EUR
FLUO022786-1G
3,4-Dichlor-3'-fluor-4'-methoxybenzophenon
3,4-Dichlor-3'-fluor-4'-methoxybenzophenon
Formel: C₁₄H₉Cl₂FO₂
MDL: MFCD06201551
CAS-Nummer: 844885-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR