3,4-Difluor-4'-methylbenzophenon

Lieferant: FLUOROCHEM

022788-1G
FLUO022788-1GEA 830 EUR
FLUO022788-1G
3,4-Difluor-4'-methylbenzophenon
3,4-Difluor-4'-methylbenzophenon
Formel: C₁₄H₁₀F₂O
MDL: MFCD01572462
CAS-Nummer: 157165-29-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR