3,4-Difluor-3'-methoxybenzophenon
Lieferant: FLUOROCHEM
022789-1G
FLUO022789-1GEA
1100
EUR
FLUO022789-1G
3,4-Difluor-3'-methoxybenzophenon
3,4-Difluor-3'-methoxybenzophenon
Formel:
C₁₄H₁₀F₂O₂ |
MDL:
MFCD06201553 CAS-Nummer: 845780-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...