3,4-Difluor-4'-methoxybenzophenon

Lieferant: FLUOROCHEM

022790-1G
FLUO022790-1GEA 1100 EUR
FLUO022790-1G
3,4-Difluor-4'-methoxybenzophenon
3,4-Difluor-4'-methoxybenzophenon
Formel: C₁₄H₁₀F₂O₂
MDL: MFCD06201554
CAS-Nummer: 845780-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR