3,4-Difluor-4'-isopropylbenzophenon

Lieferant: FLUOROCHEM

022792-1G
FLUO022792-1GEA 882 EUR
FLUO022792-1G
3,4-Difluor-4'-isopropylbenzophenon
3,4-Difluor-4'-isopropylbenzophenon
Formel: C₁₆H₁₄F₂O
MDL: MFCD06201556
CAS-Nummer: 845781-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR