3-Chlor-3',4',5-trifluorbenzophenon

Lieferant: FLUOROCHEM

022799-1G
FLUO022799-1GEA 687 EUR
FLUO022799-1G
3-Chlor-3',4',5-trifluorbenzophenon
3-Chlor-3',4',5-trifluorbenzophenon
Formel: C₁₃H₆ClF₃O
MDL: MFCD06201563
CAS-Nummer: 845781-06-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR