3,4-Difluor-3',4'-dimethoxybenzophenon

Lieferant: FLUOROCHEM

022800-1G
FLUO022800-1GEA 830 EUR
FLUO022800-1G
3,4-Difluor-3',4'-dimethoxybenzophenon
3,4-Difluor-3',4'-dimethoxybenzophenon
Formel: C₁₅H₁₂F₂O₃
MDL: MFCD06201564
CAS-Nummer: 845781-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR