3,4-Difluor-3',4'-dimethylbenzophenon

Lieferant: FLUOROCHEM

022802-1G
FLUO022802-1GEA 830 EUR
FLUO022802-1G
3,4-Difluor-3',4'-dimethylbenzophenon
3,4-Difluor-3',4'-dimethylbenzophenon
Formel: C₁₅H₁₂F₂O
MDL: MFCD03486955
CAS-Nummer: 518993-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR