3,4-Difluor-3',5'-dimethylbenzophenon
Lieferant: FLUOROCHEM
022803-1G
FLUO022803-1GEA
830
EUR
FLUO022803-1G
3,4-Difluor-3',5'-dimethylbenzophenon
3,4-Difluor-3',5'-dimethylbenzophenon
Formel:
C₁₅H₁₂F₂O |
MDL:
MFCD06201566 CAS-Nummer: 845781-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...