3,4-Difluor-4'-(ethylthio)benzophenon
Lieferant: FLUOROCHEM
022804-1G
FLUO022804-1GEA
1040
EUR
FLUO022804-1G
3,4-Difluor-4'-(ethylthio)benzophenon
3,4-Difluor-4'-(ethylthio)benzophenon
Formel:
C₁₅H₁₂F₂OS |
MDL:
MFCD06201567 CAS-Nummer: 845781-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...