4-Methoxy-3,3',4'-trifluorbenzophenon
Lieferant: FLUOROCHEM
022805-1G
FLUO022805-1GEA
830
EUR
FLUO022805-1G
4-Methoxy-3,3',4'-trifluorbenzophenon
4-Methoxy-3,3',4'-trifluorbenzophenon
Formel:
C₁₄H₉F₃O₂ |
MDL:
MFCD06201568 CAS-Nummer: 845781-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...