4-Brom-4'-butylbenzophenon

Lieferant: FLUOROCHEM

022807-1G
FLUO022807-1GEA 830 EUR
FLUO022807-1G
4-Brom-4'-butylbenzophenon
4-Brom-4'-butylbenzophenon
Formel: C₁₇H₁₇BrO
MDL: MFCD06201570
CAS-Nummer: 91404-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR