4-Brom-4'-chlorbenzophenon

Lieferant: FLUOROCHEM

Synonyme: (4-Bromophenyl)(4-chlorophenyl)methanone

022809-1G
FLUO022809-1GEA 26.7 EUR
FLUO022809-1G
4-Brom-4'-chlorbenzophenon
4-Brom-4'-chlorbenzophenon
Formel: C₁₃H₈BrClO
Molecular Weight: 295,5590 g/mol
MDL: MFCD00045208
CAS-Nummer: 27428-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR