4-Brom-3'-chlorbenzophenon

Lieferant: FLUOROCHEM

022810-1G
FLUO022810-1GEA 575 EUR
FLUO022810-1G
4-Brom-3'-chlorbenzophenon
4-Brom-3'-chlorbenzophenon
Formel: C₁₃H₈BrClO
MDL: MFCD06201571
CAS-Nummer: 27434-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR